missing translation for 'onlineSavingsMsg'
Learn More

Phenylselenenyl chloride, 98%

Catalog No. AAA1275106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1275106 5 g
AAA1275114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1275106 Supplier Thermo Scientific Chemicals Supplier No. A1275106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5707-04-0 | C6H5ClSe | 191.53 g/mol

Phenylselenenyl chloride is used in the preparation of (cyclobut-1-enylselanyl)benzene. It is an important versatile reagent used in organic synthesis. Further, it serves as a useful synthon for 2-amino alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylselenenyl chloride is used in the preparation of (cyclobut-1-enylselanyl)benzene. It is an important versatile reagent used in organic synthesis. Further, it serves as a useful synthon for 2-amino alcohols.

Solubility
Soluble in methanol.

Notes
Moisture sensitive. Incompatible with strong bases, strong oxidizing agents and strong alkalis.

Chemical Identifiers

CAS 5707-04-0
Molecular Formula C6H5ClSe
Molecular Weight (g/mol) 191.53
MDL Number MFCD00000478
InChI Key WJCXADMLESSGRI-UHFFFAOYSA-N
Synonym phenylselenenyl chloride, phenylselenyl chloride, benzeneselenenyl chloride, chloroselenobenzene, phenyl hypochloroselenoite, phenylselenium chloride, phsecl, phenylselenenylchloride, phenylselanyl chlorane, chloroselanylbenzene
PubChem CID 21928
IUPAC Name phenyl selenohypochlorite
SMILES Cl[Se]C1=CC=CC=C1

Specifications

Melting Point 60°C to 65°C
Boiling Point 120°C (20 mmHg)
Odor Stench
Quantity 5 g
UN Number UN2928
Beilstein 1237091
Sensitivity Air and moisture sensitive
Solubility Information Soluble in methanol.
Formula Weight 191.52
Percent Purity 98%
Chemical Name or Material Phenylselenenyl chloride
Show More Show Less
Hazard Category H301+H331-H373
Hazard Statement GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 227-196-2
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only