missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylselenenyl Bromide 97.0+%, TCI America™
$247.92 - $851.87
Chemical Identifiers
| CAS | 34837-55-3 |
|---|---|
| Molecular Formula | C6H5BrSe |
| Molecular Weight (g/mol) | 235.981 |
| MDL Number | MFCD00000047 |
| InChI Key | LCEFEIBEOBPPSJ-UHFFFAOYSA-N |
| Synonym | phenylselenyl bromide, benzeneselenenyl bromide, phenylselenenyl bromide, bromoselenobenzene, phsebr, phenylselenylbromide, benzeneselenyl bromide, phenylselanyl bromane, brseph, phenylseleno bromide |
| PubChem CID | 123446 |
| IUPAC Name | phenyl selenohypobromite |
| SMILES | C1=CC=C(C=C1)[Se]Br |
Chemical Identifiers
| 34837-55-3 | |
| 235.981 | |
| LCEFEIBEOBPPSJ-UHFFFAOYSA-N | |
| 123446 | |
| C1=CC=C(C=C1)[Se]Br |
| C6H5BrSe | |
| MFCD00000047 | |
| phenylselenyl bromide, benzeneselenenyl bromide, phenylselenenyl bromide, bromoselenobenzene, phsebr, phenylselenylbromide, benzeneselenyl bromide, phenylselanyl bromane, brseph, phenylseleno bromide | |
| phenyl selenohypobromite |
Specifications
| 34837-55-3 | |
| 107°C | |
| MFCD00000047 | |
| 2928 | |
| LCEFEIBEOBPPSJ-UHFFFAOYSA-N | |
| phenyl selenohypobromite | |
| 123446 | |
| ≥97.0% (T) | |
| Phenylselenenyl Bromide |
| 58°C | |
| C6H5BrSe | |
| 5 g | |
| phenylselenyl bromide, benzeneselenenyl bromide, phenylselenenyl bromide, bromoselenobenzene, phsebr, phenylselenylbromide, benzeneselenyl bromide, phenylselanyl bromane, brseph, phenylseleno bromide | |
| C1=CC=C(C=C1)[Se]Br | |
| 235.981 | |
| 235.97 | |
| Crystalline Powder |
Safety and Handling
TSCA : No