missing translation for 'onlineSavingsMsg'
Learn More

Phenylselenenyl Bromide 97.0+%, TCI America™

Catalog No. P13225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13225G 5 g
P132225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13225G Supplier TCI America Supplier No. P13225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34837-55-3
Molecular Formula C6H5BrSe
Molecular Weight (g/mol) 235.981
MDL Number MFCD00000047
InChI Key LCEFEIBEOBPPSJ-UHFFFAOYSA-N
Synonym phenylselenyl bromide, benzeneselenenyl bromide, phenylselenenyl bromide, bromoselenobenzene, phsebr, phenylselenylbromide, benzeneselenyl bromide, phenylselanyl bromane, brseph, phenylseleno bromide
PubChem CID 123446
IUPAC Name phenyl selenohypobromite
SMILES C1=CC=C(C=C1)[Se]Br

Specifications

Melting Point 58°C
Boiling Point 107°C
Quantity 5 g
UN Number 2928
Formula Weight 235.97
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylselenenyl Bromide