missing translation for 'onlineSavingsMsg'
Learn More

Phenylpropiolic Acid 98.0+%, TCI America™

Catalog No. p-7134696
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
P06105G 5 g
P061025G 25 g
2 options

Catalog No. P06105G

Supplier: TCI America P06105G

Only null left

Chemical Identifiers

CAS 637-44-5
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.15
MDL Number MFCD00004361
InChI Key XNERWVPQCYSMLC-UHFFFAOYSA-N
Synonym phenylpropiolic acid, 3-phenylpropiolic acid, phenylpropynoic acid, 3-phenylpropynoic acid, 2-propynoic acid, 3-phenyl, phenylacetylenecarboxylic acid, 3-phenyl-2-propynoic acid, phenylacetylene monocarboxylic acid, propiolic acid, 3-phenyl, phenyl propiolic acid
PubChem CID 69475
IUPAC Name 3-phenylprop-2-ynoic acid
SMILES OC(=O)C#CC1=CC=CC=C1

Specifications

Melting Point 137°C
Quantity 5 g
Formula Weight 146.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylpropiolic Acid