missing translation for 'onlineSavingsMsg'
Learn More

Phenylpropiolaldehyde diethyl acetal, 98%

Catalog No. p-7043460
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0087206 5 g
AAL0087214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0087206 Supplier Thermo Scientific Chemicals Supplier No. L0087206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6142-95-6 | C13H16O2 | 204.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6142-95-6
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.27
MDL Number MFCD00009238
InChI Key DTEGZYXCDQFSBZ-UHFFFAOYSA-N
Synonym 3,3-diethoxyprop-1-yn-1-yl benzene, phenylpropiolaldehyde diethyl acetal, phenylpropargyl aldehyde diethyl acetal, 3-phenylpropynal diethyl acetal, dehydrocinnamaldehyde-diethylacetal, benzene, 3,3-diethoxy-1-propynyl, propiolaldehyde, phenyl-, diethyl acetal, 1,1-diethoxy-3-phenylprop-2-yne, acmc-1bh6o, propiolaldehyde, diethyl acetal
PubChem CID 64818
IUPAC Name 3,3-diethoxyprop-1-ynylbenzene
SMILES CCOC(OCC)C#CC1=CC=CC=C1

Specifications

Density 0.991
Boiling Point 99°C to 100°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.517
Quantity 5 g
Beilstein 2049715
Formula Weight 204.27
Percent Purity 98%
Chemical Name or Material Phenylpropiolaldehyde diethyl acetal
EINECSNumber 228-143-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only