missing translation for 'onlineSavingsMsg'
Learn More

Phenylphosphonic Acid 98.0+%, TCI America™

Catalog No. p-7134486
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
P020425G 25 g
P0204100G 100 g
P0204500G 500 g
3 options

Catalog No. P020425G

Supplier: TCI America P020425G

Only null left

Chemical Identifiers

CAS 1571-33-1
Molecular Formula C6H7O3P
Molecular Weight (g/mol) 158.09
MDL Number MFCD00002136
InChI Key QLZHNIAADXEJJP-UHFFFAOYSA-N
Synonym benzenephosphonic acid, phosphonic acid, phenyl, phenyl-phosphonic acid, phosphonic acid, p-phenyl, phenyl phosphonic acid, unii-byd76t2868, sv7, pubchem21303, phosphonic acid,phenyl, wln: qpqo&r
PubChem CID 15295
IUPAC Name phenylphosphonic acid
SMILES OP(O)(=O)C1=CC=CC=C1

Specifications

Melting Point 163°C
Color White
Quantity 25 g
Formula Weight 158.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylphosphonic Acid
EINECSNumber (3)-2524
RTECSNumber TA0350000
TSCA Yes