missing translation for 'onlineSavingsMsg'
Learn More

Phenylphosphonic Acid 98.0+%, TCI America™

Catalog No. P020425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P020425G 25 g
P0204100G 100 g
P0204500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P020425G Supplier TCI America Supplier No. P020425G

Chemical Identifiers

CAS 1571-33-1
Molecular Formula C6H7O3P
Molecular Weight (g/mol) 158.09
MDL Number MFCD00002136
InChI Key QLZHNIAADXEJJP-UHFFFAOYSA-N
Synonym benzenephosphonic acid, phosphonic acid, phenyl, phenyl-phosphonic acid, phosphonic acid, p-phenyl, phenyl phosphonic acid, unii-byd76t2868, sv7, pubchem21303, phosphonic acid,phenyl, wln: qpqo&r
PubChem CID 15295
IUPAC Name phenylphosphonic acid
SMILES OP(O)(=O)C1=CC=CC=C1

Specifications

Melting Point 163°C
Color White
Quantity 25 g
Formula Weight 158.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylphosphonic Acid
EINECSNumber (3)-2524
RTECSNumber TA0350000
TSCA Yes