missing translation for 'onlineSavingsMsg'
Learn More

Phenylphosphonic Acid Disodium Salt Hydrate 98.0+%, TCI America™

Catalog No. P020525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P020525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P020525G Supplier TCI America Supplier No. P020525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25148-85-0
Molecular Formula C6H5Na2O3P
Molecular Weight (g/mol) 202.056
MDL Number MFCD00058972
InChI Key JOQAMSDLZYQHMX-UHFFFAOYSA-L
Synonym Benzenephosphonic Acid Disodium Salt
PubChem CID 12499978
IUPAC Name disodium;dioxido-oxo-phenyl-$l^{5}-phosphane
SMILES C1=CC=C(C=C1)P(=O)([O-])[O-].[Na+].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 202.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylphosphonic Acid Disodium Salt Hydrate
EINECSNumber (3)-2524
TSCA No