missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylphosphinic Acid 99.0+%, TCI America™
$35.41 - $247.92
Chemical Identifiers
| CAS | 1779-48-2 |
|---|---|
| Molecular Formula | C6H7O2P |
| Molecular Weight (g/mol) | 142.09 |
| MDL Number | MFCD00002131 |
| InChI Key | MLCHBQKMVKNBOV-UHFFFAOYSA-N |
| Synonym | phenylphosphinic acid, benzenephosphinic acid, benzenephosphinic acid, phosphinic acid, phenyl, phenyl phosphinic acid, phenyl phosphinic acid, benzolphosphonigs, benzolphosphonigs, benzene phosphinic acid, benzene phosphinic acid, hydroxyphenylphosphineoxide |
| PubChem CID | 6327116 |
| IUPAC Name | phenylphosphinic acid |
| SMILES | OP(=O)C1=CC=CC=C1 |
Chemical Identifiers
| 1779-48-2 | |
| 142.09 | |
| MLCHBQKMVKNBOV-UHFFFAOYSA-N | |
| 6327116 | |
| OP(=O)C1=CC=CC=C1 |
| C6H7O2P | |
| MFCD00002131 | |
| phenylphosphinic acid, benzenephosphinic acid, benzenephosphinic acid, phosphinic acid, phenyl, phenyl phosphinic acid, phenyl phosphinic acid, benzolphosphonigs, benzolphosphonigs, benzene phosphinic acid, benzene phosphinic acid, hydroxyphenylphosphineoxide | |
| phenylphosphinic acid |
Specifications
| 1779-48-2 | |
| White | |
| C6H7O2P | |
| 25 g | |
| phenylphosphinic acid, benzenephosphinic acid, benzenephosphinic acid, phosphinic acid, phenyl, phenyl phosphinic acid, phenyl phosphinic acid, benzolphosphonigs, benzolphosphonigs, benzene phosphinic acid, benzene phosphinic acid, hydroxyphenylphosphineoxide | |
| OP(=O)C1=CC=CC=C1 | |
| 142.09 | |
| 142.09 | |
| Crystalline Powder |
| 85°C | |
| 180°C | |
| MFCD00002131 | |
| 3261 | |
| MLCHBQKMVKNBOV-UHFFFAOYSA-N | |
| phenylphosphinic acid | |
| 6327116 | |
| ≥99.0% (T) | |
| Phenylphosphinic Acid |
Safety and Handling
RTECSNumber : SZ5462500
TSCA : Yes