missing translation for 'onlineSavingsMsg'
Learn More

Phenylphosphinic Acid 99.0+%, TCI America™

Catalog No. B0025500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0025500G 500 g
B002525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0025500G Supplier TCI America Supplier No. B0025500G
Only null left

Chemical Identifiers

CAS 1779-48-2
Molecular Formula C6H7O2P
Molecular Weight (g/mol) 142.09
MDL Number MFCD00002131
InChI Key MLCHBQKMVKNBOV-UHFFFAOYSA-N
Synonym phenylphosphinic acid, benzenephosphinic acid, phosphinic acid, phenyl, benzenephosphonous acid, phenyl phosphinic acid, mlchbqkmvknbov-uhfffaoysa-n, benzolphosphonigs, aure, benzene phosphinic acid, hydroxyphenylphosphineoxide, acmc-1bt5t
PubChem CID 6327116
IUPAC Name phenylphosphinic acid
SMILES OP(=O)C1=CC=CC=C1

Specifications

Melting Point 85°C
Color White
pH 1.95 (1% H2O soln.)
Boiling Point 180°C
Quantity 500 g
UN Number 3261
Formula Weight 142.09
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylphosphinic Acid
RTECSNumber SZ5462500
TSCA Yes