missing translation for 'onlineSavingsMsg'
Learn More

Phenylphosphinic acid, 98%

CAS: 1779-48-2 | C6H7O2P | 142.09 g/mol

$218.90 - $218.90

Chemical Identifiers

CAS 1779-48-2
Molecular Formula C6H7O2P
Molecular Weight (g/mol) 142.09
MDL Number MFCD00002131
InChI Key MLCHBQKMVKNBOV-UHFFFAOYSA-N
Synonym phenylphosphinic acid, acmc-1bt5t, benzenephosphinic acid, phosphinic acid, phenyl, phenyl phosphinic acid, mlchbqkmvknbov-uhfffaoysa-n, mlchbqkmvknbov-uhfffaoysa-n, aure, benzene phosphinic acid, benzene phosphinic acid, hydroxyphenylphosphineoxide
PubChem CID 6327116
IUPAC Name hydroxy-oxo-phenylphosphanium
SMILES OP(=O)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1745336
View Documents
Thermo Scientific Chemicals
A1745336
500 g
Each for $218.90
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1779-48-2
142.09
MLCHBQKMVKNBOV-UHFFFAOYSA-N
6327116
OP(=O)C1=CC=CC=C1
C6H7O2P
MFCD00002131
phenylphosphinic acid, acmc-1bt5t, benzenephosphinic acid, phosphinic acid, phenyl, phenyl phosphinic acid, mlchbqkmvknbov-uhfffaoysa-n, mlchbqkmvknbov-uhfffaoysa-n, aure, benzene phosphinic acid, benzene phosphinic acid, hydroxyphenylphosphineoxide
hydroxy-oxo-phenylphosphanium

Specifications

1779-48-2
C6H7O2P
500 g
2802244
MLCHBQKMVKNBOV-UHFFFAOYSA-N
hydroxy-oxo-phenylphosphanium
6327116
98%
83°C to 85°C
MFCD00002131
UN3261
phenylphosphinic acid, acmc-1bt5t, benzenephosphinic acid, phosphinic acid, phenyl, phenyl phosphinic acid, mlchbqkmvknbov-uhfffaoysa-n, mlchbqkmvknbov-uhfffaoysa-n, aure, benzene phosphinic acid, benzene phosphinic acid, hydroxyphenylphosphineoxide
OP(=O)C1=CC=CC=C1
142.09
142.1
Phenylphosphinic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335-H500

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 217-217-3

RTECSNumber : SZ5462500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only