missing translation for 'onlineSavingsMsg'
Learn More

Phenylphosphinic acid, 98%

Catalog No. AAA1745336
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1745336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1745336 Supplier Thermo Scientific Chemicals Supplier No. A1745336
Only null left

CAS: 1779-48-2 | C6H7O2P | 142.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1779-48-2
Molecular Formula C6H7O2P
Molecular Weight (g/mol) 142.09
MDL Number MFCD00002131
InChI Key MLCHBQKMVKNBOV-UHFFFAOYSA-N
Synonym phenylphosphinic acid, benzenephosphinic acid, phosphinic acid, phenyl, benzenephosphonous acid, phenyl phosphinic acid, mlchbqkmvknbov-uhfffaoysa-n, benzolphosphonigs, aure, benzene phosphinic acid, hydroxyphenylphosphineoxide, acmc-1bt5t
PubChem CID 6327116
IUPAC Name hydroxy-oxo-phenylphosphanium
SMILES OP(=O)C1=CC=CC=C1

Specifications

Melting Point 83°C to 85°C
Quantity 500 g
UN Number UN3261
Beilstein 2802244
Formula Weight 142.1
Percent Purity 98%
Chemical Name or Material Phenylphosphinic acid
Hazard Category H302-H314-H335-H500
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 217-217-3
RTECSNumber SZ5462500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only