missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylmethylsulfonyl Fluoride Crystalline MP Biomedicals
Supplier: MP Biomedicals Inc 0219538105
Specifications
| Phenylmethylsulfonyl Fluoride | |
| C7H7FO2S | |
| phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride | |
| C1=CC=C(C=C1)CS(=O)(=O)F | |
| 174.189 | |
| CHEBI:8102 | |
| Crystalline |
| 329-98-6 | |
| 5 g | |
| YBYRMVIVWMBXKQ-UHFFFAOYSA-N | |
| phenylmethanesulfonyl fluoride | |
| 4784 | |
| 174.2 |
Chemical Identifiers
| 329-98-6 | |
| 174.189 | |
| phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride | |
| CHEBI:8102 | |
| C1=CC=C(C=C1)CS(=O)(=O)F |
| C7H7FO2S | |
| YBYRMVIVWMBXKQ-UHFFFAOYSA-N | |
| 4784 | |
| phenylmethanesulfonyl fluoride |