Learn More
Thermo Scientific Chemicals Phenylmethanesulfonyl fluoride, 99%
CAS: 329-98-6 | C7H7FO2S | 174.19 g/mol
Supplier: Thermo Scientific Chemicals 215740050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Phenylmethanesulfonyl fluoride | |
| 98.5 | |
| 90.0°C to 94.0°C | |
| Authentic | |
| Glass bottle | |
| C6H5CH2SO2F | |
| 5 g | |
| 15, 7659 | |
| Solubility in water: hydrolysis. Other solubilities: soluble in xylene and petroleum ether | |
| C1=CC=C(C=C1)CS(=O)(=O)F | |
| 174.19 | |
| CHEBI:8102 | |
| 99% |
| 329-98-6 | |
| 100.0 | |
| White | |
| 98.5% min. (GC) | |
| C7H7FO2S | |
| MFCD00007424 | |
| 11, III, 331 | |
| phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride | |
| YBYRMVIVWMBXKQ-UHFFFAOYSA-N | |
| phenylmethanesulfonyl fluoride | |
| 4784 | |
| 174.19 | |
| Needles (May Agglomerate) |
Chemical Identifiers
| 329-98-6 | |
| 174.19 | |
| YBYRMVIVWMBXKQ-UHFFFAOYSA-N | |
| 4784 | |
| phenylmethanesulfonyl fluoride |
| C7H7FO2S | |
| MFCD00007424 | |
| phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride | |
| CHEBI:8102 | |
| C1=CC=C(C=C1)CS(=O)(=O)F |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minut
GHS Signal Word: Danger
EINECSNumber : 206-350-2
RUO – Research Use Only