missing translation for 'onlineSavingsMsg'
Learn More

Phenylmalonic Acid 95.0+%, TCI America™

Catalog No. P098725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P098725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P098725G Supplier TCI America Supplier No. P098725G

Specifications

CAS 2613-89-0
Color White
Molecular Formula C9H8O4
Quantity 25 g
MDL Number MFCD00004253
Synonym phenylmalonic acid, 2-phenylmalonic acid, propanedioic acid, phenyl, phenylpropanedioc acid, phenylmalonicacid, malonic acid, phenyl, chembl78794, propanedioic acid, 2-phenyl, .alpha.-carboxyphenylacetic acid, toluene-.alpha.,.alpha.-dicarboxylic acid
InChI Key WWYDYZMNFQIYPT-UHFFFAOYSA-N
SMILES OC(=O)C(C(O)=O)C1=CC=CC=C1
IUPAC Name 2-phenylpropanedioic acid
Molecular Weight (g/mol) 180.16
PubChem CID 75791
Formula Weight 180.16
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylmalonic Acid
Show More Show Less