missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylmaleic Anhydride 98.0+%, TCI America™
$167.76 - $574.13
Chemical Identifiers
| CAS | 36122-35-7 |
|---|---|
| Molecular Formula | C10H6O3 |
| Molecular Weight (g/mol) | 174.16 |
| MDL Number | MFCD00015472 |
| InChI Key | QZYCWJVSPFQUQC-UHFFFAOYSA-N |
| Synonym | phenylmaleic anhydride, 3-phenyl-furan-2,5-dione, 3-phenyl-2,5-furandione, 3-phenyl-2,5-dihydrofuran-2,5-dione, 2,5-furandione, 3-phenyl, phenylmaleicanhydride, phenylmalecanhydrde, acmc-20ajwi, phenyl maleic anhydride, phenyl-maleic anhydride |
| PubChem CID | 99174 |
| IUPAC Name | 3-phenyl-2,5-dihydrofuran-2,5-dione |
| SMILES | O=C1OC(=O)C(=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 36122-35-7 | |
| 174.16 | |
| QZYCWJVSPFQUQC-UHFFFAOYSA-N | |
| 99174 | |
| O=C1OC(=O)C(=C1)C1=CC=CC=C1 |
| C10H6O3 | |
| MFCD00015472 | |
| phenylmaleic anhydride, 3-phenyl-furan-2,5-dione, 3-phenyl-2,5-furandione, 3-phenyl-2,5-dihydrofuran-2,5-dione, 2,5-furandione, 3-phenyl, phenylmaleicanhydride, phenylmalecanhydrde, acmc-20ajwi, phenyl maleic anhydride, phenyl-maleic anhydride | |
| 3-phenyl-2,5-dihydrofuran-2,5-dione |
Specifications
| 36122-35-7 | |
| White-Yellow | |
| MFCD00015472 | |
| phenylmaleic anhydride, 3-phenyl-furan-2,5-dione, 3-phenyl-2,5-furandione, 3-phenyl-2,5-dihydrofuran-2,5-dione, 2,5-furandione, 3-phenyl, phenylmaleicanhydride, phenylmalecanhydrde, acmc-20ajwi, phenyl maleic anhydride, phenyl-maleic anhydride | |
| O=C1OC(=O)C(=C1)C1=CC=CC=C1 | |
| 174.16 | |
| 174.16 | |
| Crystalline Powder |
| 122°C | |
| C10H6O3 | |
| 1 g | |
| QZYCWJVSPFQUQC-UHFFFAOYSA-N | |
| 3-phenyl-2,5-dihydrofuran-2,5-dione | |
| 99174 | |
| ≥98.0% (GC,T) | |
| Phenylmaleic Anhydride |
Safety and Handling
EINECSNumber : (3)-3109
TSCA : No