Learn More
Phenylmaleic anhydride, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 310400100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Phenylmaleic anhydride | |
| 122.0°C to 125.0°C (0.2 Torr) | |
| 99% | |
| C10H6O3 | |
| 10g | |
| QZYCWJVSPFQUQC-UHFFFAOYSA-N | |
| 3-phenylfuran-2,5-dione | |
| 99174 | |
| 99% |
| 36122-35-7 | |
| Authentic | |
| Glass bottle | |
| MFCD00015472 | |
| phenylmaleic anhydride, 3-phenyl-furan-2,5-dione, 3-phenyl-2,5-furandione, 3-phenyl-2,5-dihydrofuran-2,5-dione, 2,5-furandione, 3-phenyl, phenylmaleicanhydride, phenylmalecanhydrde, acmc-20ajwi, phenyl maleic anhydride, phenyl-maleic anhydride | |
| O=C1OC(=O)C(=C1)C1=CC=CC=C1 | |
| 174.16 | |
| 174.16 |
Chemical Identifiers
| 36122-35-7 | |
| 174.16 | |
| QZYCWJVSPFQUQC-UHFFFAOYSA-N | |
| 99174 | |
| O=C1OC(=O)C(=C1)C1=CC=CC=C1 |
| C10H6O3 | |
| MFCD00015472 | |
| phenylmaleic anhydride, 3-phenyl-furan-2,5-dione, 3-phenyl-2,5-furandione, 3-phenyl-2,5-dihydrofuran-2,5-dione, 2,5-furandione, 3-phenyl, phenylmaleicanhydride, phenylmalecanhydrde, acmc-20ajwi, phenyl maleic anhydride, phenyl-maleic anhydride | |
| 3-phenylfuran-2,5-dione |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 252-881-8
RUO â Research Use Only