missing translation for 'onlineSavingsMsg'
Learn More

Phenylmaleic anhydride, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 310400010

 View more versions of this product

Catalog No. AC310400010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Phenylmaleic anhydride
122.0°C to 125.0°C (0.2 Torr)
99%
C10H6O3
1g
QZYCWJVSPFQUQC-UHFFFAOYSA-N
3-phenylfuran-2,5-dione
99174
99%
36122-35-7
Authentic
Glass bottle
MFCD00015472
phenylmaleic anhydride, 3-phenyl-furan-2,5-dione, 3-phenyl-2,5-furandione, 3-phenyl-2,5-dihydrofuran-2,5-dione, 2,5-furandione, 3-phenyl, phenylmaleicanhydride, phenylmalecanhydrde, acmc-20ajwi, phenyl maleic anhydride, phenyl-maleic anhydride
O=C1OC(=O)C(=C1)C1=CC=CC=C1
174.16
174.16

Chemical Identifiers

36122-35-7
174.16
QZYCWJVSPFQUQC-UHFFFAOYSA-N
99174
O=C1OC(=O)C(=C1)C1=CC=CC=C1
C10H6O3
MFCD00015472
phenylmaleic anhydride, 3-phenyl-furan-2,5-dione, 3-phenyl-2,5-furandione, 3-phenyl-2,5-dihydrofuran-2,5-dione, 2,5-furandione, 3-phenyl, phenylmaleicanhydride, phenylmalecanhydrde, acmc-20ajwi, phenyl maleic anhydride, phenyl-maleic anhydride
3-phenylfuran-2,5-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 252-881-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only