missing translation for 'onlineSavingsMsg'
Learn More

Phenylglyoxal monohydrate, 97%

CAS: 1075-06-5 | C8H6O2·H2O | 152.15 g/mol

$166.35 - $166.35

Chemical Identifiers

CAS 1075-06-5
Molecular Formula C8H6O2·H2O
Molecular Weight (g/mol) 152.15
MDL Number MFCD00149499
InChI Key NBIBDIKAOBCFJN-UHFFFAOYSA-N
Synonym dihydroxyacetophenone, acetophenone, dihydroxy, dihydroxy 1-phenylethanone, 2,2-dihydroxy-1-phenyl-ethanone, ethanone, 2,2-dihydroxy-1-phenyl, acetophenone, 2,2-dihydroxy, alpha,alpha-dihydroxyacetophenone, dioxyacetophenone, alpha,alpha-dihydroxy-acetophenone
PubChem CID 99611
IUPAC Name 2,2-dihydroxy-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC161970050
SDS View Documents
Thermo Scientific Chemicals
161970050
5 g Glass bottle
Each for $166.35
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1075-06-5
152.15
NBIBDIKAOBCFJN-UHFFFAOYSA-N
99611
C1=CC=C(C=C1)C(=O)C(O)O
C8H6O2·H2O
MFCD00149499
dihydroxyacetophenone, acetophenone, dihydroxy, dihydroxy 1-phenylethanone, 2,2-dihydroxy-1-phenyl-ethanone, ethanone, 2,2-dihydroxy-1-phenyl, acetophenone, 2,2-dihydroxy, alpha,alpha-dihydroxyacetophenone, dioxyacetophenone, alpha,alpha-dihydroxy-acetophenone
2,2-dihydroxy-1-phenylethanone

Specifications

1075-06-5
Pink or White to Yellow
Authentic
Glass bottle
C6H5COCHO·H2O
5 g
dihydroxyacetophenone, acetophenone, dihydroxy, dihydroxy 1-phenylethanone, 2,2-dihydroxy-1-phenyl-ethanone, ethanone, 2,2-dihydroxy-1-phenyl, acetophenone, 2,2-dihydroxy, alpha,alpha-dihydroxyacetophenone, dioxyacetophenone, alpha,alpha-dihydroxy-acetophenone
C1=CC=C(C=C1)C(=O)C(O)O
152.15
152.15
Powder
71.0°C to 76.0°C
142.0°C (125.0 mmHg)
97%
C8H6O2·H2O
MFCD00149499
07, 670
NBIBDIKAOBCFJN-UHFFFAOYSA-N
2,2-dihydroxy-1-phenylethanone
99611
97%
Phenylglyoxal monohydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only