missing translation for 'onlineSavingsMsg'
Learn More

Phenylglyoxal monohydrate, 97%

Catalog No. AC161971000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161971000 100 g Glass bottle
AC161970050 5 g Glass bottle
AC161970250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC161971000 Supplier Thermo Scientific Chemicals Supplier No. 161971000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1075-06-5 | C8H6O2·H2O | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1075-06-5
Molecular Formula C8H6O2·H2O
Molecular Weight (g/mol) 152.15
MDL Number MFCD00149499
InChI Key NBIBDIKAOBCFJN-UHFFFAOYSA-N
Synonym dihydroxyacetophenone, acetophenone, dihydroxy, dihydroxy 1-phenylethanone, 2,2-dihydroxy-1-phenyl-ethanone, ethanone, 2,2-dihydroxy-1-phenyl, acetophenone, 2,2-dihydroxy, alpha,alpha-dihydroxyacetophenone, dioxyacetophenone, alpha,alpha-dihydroxy-acetophenone
PubChem CID 99611
IUPAC Name 2,2-dihydroxy-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(O)O

Specifications

Melting Point 71.0°C to 76.0°C
Color Pink or White to Yellow
Boiling Point 142.0°C (125.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5COCHO·H2O
Quantity 100 g
Beilstein 07, 670
Formula Weight 152.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Phenylglyoxal monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only