missing translation for 'onlineSavingsMsg'
Learn More

Phenylglyoxal monohydrate, 97%

CAS: 1075-06-5 | C8H8O3 | 152.149 g/mol

Supplier:  Thermo Scientific Chemicals A1067714

 View more versions of this product

Catalog No. AAA1067714


Only null left
Explore available promotions

Description

Description

Phenylglyoxal monohydrate was used to modify pig muscle carbonic anhydraseIII and as a specific reagent for arginine groups. It was also used to prepare pyrrolinone and furan derivatives and as as a chemiluminescent reagent for the determination of purines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylglyoxal monohydrate was used to modify pig muscle carbonic anhydraseIII and as a specific reagent for arginine groups. It was also used to prepare pyrrolinone and furan derivatives and as as a chemiluminescent reagent for the determination of purines.

Solubility
Partly miscible with water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Phenylglyoxal monohydrate
71°C to 77°C
C8H8O3
25 g
dihydroxyacetophenone, acetophenone, dihydroxy, dihydroxy 1-phenylethanone, 2,2-dihydroxy-1-phenyl-ethanone, ethanone, 2,2-dihydroxy-1-phenyl, acetophenone, 2,2-dihydroxy, alpha,alpha-dihydroxyacetophenone, dioxyacetophenone, alpha,alpha-dihydroxy-acetophenone
NBIBDIKAOBCFJN-UHFFFAOYSA-N
2,2-dihydroxy-1-phenylethanone
99611
97%
1075-06-5
142°C (125 mmHg)
MFCD00149499
1854721
Partly miscible with water.
C1=CC=C(C=C1)C(=O)C(O)O
152.149
152.15 (134.12 Anhydrous)

Chemical Identifiers

1075-06-5
152.149
NBIBDIKAOBCFJN-UHFFFAOYSA-N
99611
C1=CC=C(C=C1)C(=O)C(O)O
C8H8O3
MFCD00149499
dihydroxyacetophenone, acetophenone, dihydroxy, dihydroxy 1-phenylethanone, 2,2-dihydroxy-1-phenyl-ethanone, ethanone, 2,2-dihydroxy-1-phenyl, acetophenone, 2,2-dihydroxy, alpha,alpha-dihydroxyacetophenone, dioxyacetophenone, alpha,alpha-dihydroxy-acetophenone
2,2-dihydroxy-1-phenylethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 214-036-1

RTECSNumber : KM5775180

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only