missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylfluorone 97.0+%, TCI America™
$67.38 - $190.59
Chemical Identifiers
| CAS | 975-17-7 |
|---|---|
| Molecular Formula | C19H12O5 |
| Molecular Weight (g/mol) | 320.3 |
| MDL Number | MFCD00005048 |
| InChI Key | YDCFOUBAMGLLKA-UHFFFAOYSA-N |
| Synonym | phenylfluorone, fluorone black, 9-phenyl-2,3,7-trihydroxy-6-fluorone, fluorone, phenyl, 2,6,7-trihydroxy-9-phenyl-3h-xanthen-3-one, 9-pheny-3-fluorone, 3h-xanthen-3-one, 2,6,7-trihydroxy-9-phenyl, 9-phenyl-3-fluorone, 2,3,7-trihydroxy-9-phenyl-6-fluorone, 2,6,7-trihydroxy-9-phenyl-3-isoxanthone |
| PubChem CID | 70420 |
| IUPAC Name | 2,6,7-trihydroxy-9-phenylxanthen-3-one |
| SMILES | C1=CC=C(C=C1)C2=C3C=C(C(=O)C=C3OC4=CC(=C(C=C42)O)O)O |
Chemical Identifiers
| 975-17-7 | |
| 320.3 | |
| YDCFOUBAMGLLKA-UHFFFAOYSA-N | |
| 70420 | |
| C1=CC=C(C=C1)C2=C3C=C(C(=O)C=C3OC4=CC(=C(C=C42)O)O)O |
| C19H12O5 | |
| MFCD00005048 | |
| phenylfluorone, fluorone black, 9-phenyl-2,3,7-trihydroxy-6-fluorone, fluorone, phenyl, 2,6,7-trihydroxy-9-phenyl-3h-xanthen-3-one, 9-pheny-3-fluorone, 3h-xanthen-3-one, 2,6,7-trihydroxy-9-phenyl, 9-phenyl-3-fluorone, 2,3,7-trihydroxy-9-phenyl-6-fluorone, 2,6,7-trihydroxy-9-phenyl-3-isoxanthone | |
| 2,6,7-trihydroxy-9-phenylxanthen-3-one |
Specifications
| 975-17-7 | |
| MFCD00005048 | |
| phenylfluorone, fluorone black, 9-phenyl-2,3,7-trihydroxy-6-fluorone, fluorone, phenyl, 2,6,7-trihydroxy-9-phenyl-3h-xanthen-3-one, 9-pheny-3-fluorone, 3h-xanthen-3-one, 2,6,7-trihydroxy-9-phenyl, 9-phenyl-3-fluorone, 2,3,7-trihydroxy-9-phenyl-6-fluorone, 2,6,7-trihydroxy-9-phenyl-3-isoxanthone | |
| C1=CC=C(C=C1)C2=C3C=C(C(=O)C=C3OC4=CC(=C(C=C42)O)O)O | |
| 320.3 | |
| 320.30 | |
| Crystalline Powder |
| C19H12O5 | |
| 1 g | |
| YDCFOUBAMGLLKA-UHFFFAOYSA-N | |
| 2,6,7-trihydroxy-9-phenylxanthen-3-one | |
| 70420 | |
| ≥97.0% (HPLC) | |
| Phenylfluorone |
Safety and Handling
TSCA : Yes