missing translation for 'onlineSavingsMsg'
Learn More

Phenylboronic acid pinacol ester, 97%

Catalog No. AC445220250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC445220250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC445220250 Supplier Thermo Scientific Chemicals Supplier No. 445220250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24388-23-6 | C12H17BO2 | 204.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24388-23-6
Molecular Formula C12H17BO2
Molecular Weight (g/mol) 204.08
MDL Number MFCD00966985 MFCD31699955
InChI Key KKLCYBZPQDOFQK-UHFFFAOYSA-N
Synonym phenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene, phenylboronic acid, pinacol ester, 2-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzeneboronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-phenyl, acmc-209gbq, phenylboronic pinacol ester, phenylboronicacidpinacolester, pinacolboryl benzene
PubChem CID 570673
IUPAC Name 4,4,5,5-tetramethyl-2-phenyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC=C1

Specifications

Melting Point 27°C to 31°C
Boiling Point 110°C (12.0 Torr)
Flash Point 107°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 204.08
Percent Purity 97%
Chemical Name or Material Phenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only