missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Phenylbis(2,4,6-trimethylbenzoyl)phosphine oxide
Description
Phenylbis(2,4,6-trimethylbenzoyl)phosphine oxide
Specifications
Specifications
| CAS | 162881-26-7 |
| Melting Point | 131°C to 135°C |
| Molecular Formula | C26H27O3P |
| Linear Formula | [(CH3)3C6H2CO]2 P(O)C6H5 |
| MDL Number | MFCD01863675 |
| Quantity | 50 g |
| Synonym | BAPOs; Bisacylphosphine oxides |
| Molecular Weight (g/mol) | 418.46 |
| Percent Purity | 97% |