missing translation for 'onlineSavingsMsg'
Learn More

Phenylbiguanide 98.0+%, TCI America™

Catalog No. P100210G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P100210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P100210G Supplier TCI America Supplier No. P100210G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 102-02-3
Molecular Formula C8H11N5
Molecular Weight (g/mol) 177.211
MDL Number MFCD00179077
InChI Key CUQCMXFWIMOWRP-UHFFFAOYSA-N
Synonym phenylbiguanide, 1-phenylbiguanide, phenyl biguanide, phenyldiguanide, phenylguanide, phenyl diguanide, n-phenyl-n'-guanylguanidine, n-phenylimidodicarbonimidic diamide, biguanide, phenyl, imidodicarbonimidic diamide, n-phenyl
PubChem CID 4780
ChEBI CHEBI:75377
IUPAC Name 1-(diaminomethylidene)-2-phenylguanidine
SMILES C1=CC=C(C=C1)N=C(N)N=C(N)N

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 10 g
Formula Weight 177.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylbiguanide
EINECSNumber (3)-3212
RTECSNumber DU2450000
TSCA No