missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsine Oxide, Reagent Grade, LabChem™

Catalog No. LC183357
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC183357 10 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC183357 Supplier LabChem Supplier No. LC183357

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 637-03-6
Molecular Formula C6H5AsO
Molecular Weight (g/mol) 168.03
MDL Number MFCD00001990
InChI Key BQVCCPGCDUSGOE-UHFFFAOYSA-N
Synonym phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
PubChem CID 4778
ChEBI CHEBI:75253
IUPAC Name arsorosobenzene
SMILES O=[As]C1=CC=CC=C1

Specifications

CAS Max % 100
Melting Point 145°C
Color White
Packaging Amber Glass
Linear Formula C6H5AsO
Quantity 10 g
UN Number UN1557
Solubility Information Soluble in water
Formula Weight 168.03
Grade Reagent
Physical Form Powder
Chemical Name or Material Phenylarsine Oxide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Toxic if swallowed or if inhaled.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid breathing dust.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Avoid release to the environment.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 211-275-3
Recommended Storage Room Temperature