missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsine Oxide (PAO) Titrant, 0.0250 N (N/40), Stabilized, Ricca Chemical

Catalog No. 57551
Change view
Click to view available options
Package: Amber Glass; Size: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
57551 Package: Amber Glass; Size: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57551 Supplier Ricca Chemical Company Supplier No. 57551

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 637-03-6, 7732-18-5, 1310-73-2, 7732-18-5
CAS Min % 0.25, 98.0, 0.1
CAS Max % 0.26, 100.0, 0.1
Molecular Formula C6H5AsO
MDL Number MFCD00001990
Synonym phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
InChI Key BQVCCPGCDUSGOE-UHFFFAOYSA-N
SMILES O=[As]C1=CC=CC=C1
IUPAC Name arsorosobenzene
Molecular Weight (g/mol) 168.03
PubChem CID 4778  
ChEBI CHEBI:75253
Packaging Amber Glass Bottle
Treatment(s) Stabilized
Quantity 4 L
Concentration or Composition Notes 0.0250Normal
Chemical Name or Material Phenylarsine Oxide (PAO) Titrant
Show More Show Less