missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylarsine oxide, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 145°C to 148°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97% |
| Packaging | Glass bottle |
| Linear Formula | C6H5As=O |
| Quantity | 1 g |
| Beilstein | 16, I, 438 |
| Merck Index | 15, 7048 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: very soluble in benzene,slightly soluble in ethanol,insoluble in ether |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if inhaled. Very toxic to aquatic life with long lasting effects. Toxic if swallowed. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Avoid release to the environment. |
| EINECSNumber | 211-275-3 |
RUO – Research Use Only