missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsine oxide, 97%

Catalog No. AAA1467403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1467403 1 g
AAA1467406 5 g
AAA1467414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1467403 Supplier Thermo Scientific Chemicals Supplier No. A1467403
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 637-03-6 | C6H5AsO | 168.03 g/mol

Phenylarsine oxide is used as a poultry coccidiostat in veterinary medicine. It is also used in a membrane-permeable protein tyrosine phosphatase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylarsine oxide is used as a poultry coccidiostat in veterinary medicine. It is also used in a membrane-permeable protein tyrosine phosphatase inhibitor.

Solubility
Soluble in dimethyl sulfoxide, chloroform, benzene, ether, ethanol, 50:50 dimethyl sulfoxide:1 M hydrochloric acid (3.0 mg/ml) and toluene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 637-03-6
Molecular Formula C6H5AsO
Molecular Weight (g/mol) 168.03
MDL Number MFCD00001990
InChI Key BQVCCPGCDUSGOE-UHFFFAOYSA-N
Synonym phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
PubChem CID 4778
ChEBI CHEBI:75253
IUPAC Name arsorosobenzene
SMILES O=[As]C1=CC=CC=C1

Specifications

Melting Point 140°C to 160°C
Quantity 1 g
UN Number UN3465
Beilstein 2935227
Merck Index 14,6949
Solubility Information Soluble in dimethyl sulfoxide,chloroform,benzene,ether,ethanol,50:50 dimethyl sulfoxide:1 M hydrochloric acid (3.0mg/ml) and toluene. Insoluble in water.
Formula Weight 168.03
Percent Purity 97%
Chemical Name or Material Phenylarsine oxide
Hazard Category H301+H331
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P261-P264-P270-P271-P301+P310-P304+P340-P311-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 211-275-3
RTECSNumber CH8100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only