missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetyl Chloride 98.0+%, TCI America™

Catalog No. p-7134437
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P012925G 25 g
P0129500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P012925G Supplier TCI America Supplier No. P012925G
Only null left

Chemical Identifiers

CAS 103-80-0
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000729
InChI Key VMZCDNSFRSVYKQ-UHFFFAOYSA-N
Synonym phenylacetyl chloride, benzeneacetyl chloride, acetyl chloride, phenyl, phenylacetic acid chloride, phenacetyl chloride, phenylacetylchloride, unii-t30899drnd, phenyl acetyl chloride, a-toluoyl chloride, phenylacetyl
PubChem CID 7679
IUPAC Name 2-phenylacetyl chloride
SMILES ClC(=O)CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 211°C
Quantity 25 g
UN Number 2577
Formula Weight 154.59
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Phenylacetyl Chloride
EINECSNumber (3)-1722
TSCA Yes