missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetaldehyde Dimethyl Acetal 97.0+%, TCI America™

Catalog No. P1161500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1161500ML 500 mL
P116125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1161500ML Supplier TCI America Supplier No. P1161500ML

Chemical Identifiers

CAS 101-48-4
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00008487
InChI Key WNJSKZBEWNVKGU-UHFFFAOYSA-N
Synonym 2,2-dimethoxyethyl benzene, phenylacetaldehyde dimethyl acetal, viridine, hyscylene p, 1,1-dimethoxy-2-phenylethane, benzene, 2,2-dimethoxyethyl, phenacetaldehyde dimethyl acetal, padma, 2,2-dimethoxy-1-phenylethane, phenylacetic aldehyde dimethyl acetal
PubChem CID 60995
IUPAC Name 2,2-dimethoxyethylbenzene
SMILES COC(CC1=CC=CC=C1)OC

Specifications

Color Yellow
Boiling Point 108°C
Quantity 500 mL
Formula Weight 166.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenylacetaldehyde Dimethyl Acetal
EINECSNumber (3)-1153
RTECSNumber AB3040000
TSCA Yes