Learn More
Phenyl vinyl sulfone, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 221250250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Phenyl vinyl sulfone | |
| Authentic | |
| C8H8O2S | |
| MFCD00007554 | |
| 10,315; 13,335 | |
| phenyl vinyl sulfone, vinylsulfonyl benzene, ethenesulfonyl benzene, benzene, ethenylsulfonyl, sulfone, phenyl vinyl, vinylsulfonylbenzene, phenyl vinyl sulphone, ethenylsulfonyl benzene, ethenesulfonyl-benzene, phenyl-vinylsulfon | |
| UJTPZISIAWDGFF-UHFFFAOYSA-N | |
| ethenylsulfonylbenzene | |
| 79664 | |
| 98% |
| 5535-48-8 | |
| 97.5% min. (GC) | |
| C6H5SO2CH=CH2 | |
| 25g | |
| 15,7431 | |
| Solubility in water: slightly soluble. | |
| C=CS(=O)(=O)C1=CC=CC=C1 | |
| 168.21 | |
| 168.21 |
Chemical Identifiers
| 5535-48-8 | |
| 168.21 | |
| UJTPZISIAWDGFF-UHFFFAOYSA-N | |
| 79664 | |
| C=CS(=O)(=O)C1=CC=CC=C1 |
| C8H8O2S | |
| MFCD00007554 | |
| phenyl vinyl sulfone, vinylsulfonyl benzene, ethenesulfonyl benzene, benzene, ethenylsulfonyl, sulfone, phenyl vinyl, vinylsulfonylbenzene, phenyl vinyl sulphone, ethenylsulfonyl benzene, ethenesulfonyl-benzene, phenyl-vinylsulfon | |
| ethenylsulfonylbenzene |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
GHS Signal Word: Danger
EINECSNumber : 226-890-2
RUO â Research Use Only