Learn More
Phenyl selenocyanate, 98%
CAS: 2179-79-5 | C7H5NSe | 182.095 g/mol
Supplier: Thermo Scientific Chemicals L2019406
Description
Phenyl selenocyanate is used as a specific source of nucleophilic and electrophilic selenium species in synthetic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenyl selenocyanate is used as a specific source of nucleophilic and electrophilic selenium species in synthetic chemistry.
Solubility
Miscible with organic solvents, terahydro furan, dichloromethane and acetonitrile.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
| Phenyl selenocyanate | |
| 1.484 | |
| >110°C (230°F) | |
| 1.603 | |
| 5 g | |
| 1617370 | |
| Miscible with organic solvents,terahydro furan,dichloromethane and acetonitrile. | |
| C1=CC=C(C=C1)[Se]C#N | |
| 182.095 | |
| 182.08 |
| 2179-79-5 | |
| 116°C to 117°C (12 mmHg) | |
| C7H5NSe | |
| MFCD00216944 | |
| UN3440 | |
| phenylselenocyanate, phenylselanyl formonitrile, selenocyanic acid, phenyl ester, acmc-1cgab, phenyl selenocyanate, selenocyanic acid phenyl ester, 2-phenyl-2-selenaethanenitrile | |
| NODWRXQVQYOJGN-UHFFFAOYSA-N | |
| phenyl selenocyanate | |
| 555340 | |
| 98% |
Chemical Identifiers
| 2179-79-5 | |
| 182.095 | |
| NODWRXQVQYOJGN-UHFFFAOYSA-N | |
| 555340 | |
| C1=CC=C(C=C1)[Se]C#N |
| C7H5NSe | |
| MFCD00216944 | |
| phenylselenocyanate, phenylselanyl formonitrile, selenocyanic acid, phenyl ester, acmc-1cgab, phenyl selenocyanate, selenocyanic acid phenyl ester, 2-phenyl-2-selenaethanenitrile | |
| phenyl selenocyanate |
Safety and Handling
GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
H301-H330-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, LIQUID, N.O.S.
TSCA : No
RUO – Research Use Only