missing translation for 'onlineSavingsMsg'
Learn More

Phenyl selenocyanate, 98%

Catalog No. AAL2019403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2019403 1 g
AAL2019406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2019403 Supplier Thermo Scientific Chemicals Supplier No. L2019403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2179-79-5 | C7H5NSe | 182.095 g/mol

Phenyl selenocyanate is used as a specific source of nucleophilic and electrophilic selenium species in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl selenocyanate is used as a specific source of nucleophilic and electrophilic selenium species in synthetic chemistry.

Solubility
Miscible with organic solvents, terahydro furan, dichloromethane and acetonitrile.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2179-79-5
Molecular Formula C7H5NSe
Molecular Weight (g/mol) 182.095
MDL Number MFCD00216944
InChI Key NODWRXQVQYOJGN-UHFFFAOYSA-N
Synonym phenylselenocyanate, phenylselanyl formonitrile, selenocyanic acid, phenyl ester, acmc-1cgab, phenyl selenocyanate, selenocyanic acid phenyl ester, 2-phenyl-2-selenaethanenitrile
PubChem CID 555340
IUPAC Name phenyl selenocyanate
SMILES C1=CC=C(C=C1)[Se]C#N

Specifications

Density 1.484
Boiling Point 116°C to 117°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.603
Quantity 1 g
UN Number UN3440
Beilstein 1617370
Solubility Information Miscible with organic solvents,terahydro furan,dichloromethane and acetonitrile.
Formula Weight 182.08
Percent Purity 98%
Chemical Name or Material Phenyl selenocyanate
Show More Show Less
Hazard Category H301-H330-H373
Hazard Statement GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, LIQUID, N.O.S.
TSCA No

RUO – Research Use Only