missing translation for 'onlineSavingsMsg'
Learn More

Phenyl propargyl ether, 98+%

Catalog No. aaa1827206
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1827206 5 g
AAA1827203 1 g
AAA1827214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1827206 Supplier Thermo Scientific Chemicals Supplier No. A1827206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13610-02-1 | C9H8O | 132.16 g/mol

Phenyl propargyl ether is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl propargyl ether is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water (1.0 g/L at 25°C).

Notes
Air and light sensitive. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 13610-02-1
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00048107
InChI Key AIQRJSXKXVZCJO-UHFFFAOYSA-N
Synonym phenyl propargyl ether, prop-2-ynyloxy benzene, prop-2-yn-1-yloxy benzene, phenyl 2-propynyl ether, benzene, 2-propynyloxy, 3-phenoxy-1-propyne, 2-propynyloxy benzene, phenylpropargyl ether, prop-2-ynyloxybenzene, ether, phenyl 2-propynyl
PubChem CID 83613
IUPAC Name prop-2-ynoxybenzene
SMILES C#CCOC1=CC=CC=C1

Specifications

Density 1.03
Boiling Point 89°C to 90°C (14 mmHg)
Flash Point 76°C (169°F)
Odor Pungent
Refractive Index 1.533
Linear Formula C6H5OCH2C≡CH
Quantity 5 g
Sensitivity Air and light sensitive
Solubility Information Slightly soluble in water (1.0g/L at 25°C).
Formula Weight 132.16
Percent Purity ≥98%
Chemical Name or Material Phenyl propargyl ether
Show More Show Less
Hazard Category H227-H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H227-H315
Causes serious eye damage.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 237-095-5
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only