missing translation for 'onlineSavingsMsg'
Learn More

Phenyl propargyl ether, 97+%, Thermo Scientific™

Catalog No. p-3598451
Change view
Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392730050 5mL Glass bottle
AC392730250 25mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC392730050 Supplier Thermo Scientific Chemicals Supplier No. 392730050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 13610-02-1
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00048107
InChI Key AIQRJSXKXVZCJO-UHFFFAOYSA-N
Synonym phenyl propargyl ether, prop-2-ynyloxy benzene, prop-2-yn-1-yloxy benzene, phenyl 2-propynyl ether, benzene, 2-propynyloxy, 3-phenoxy-1-propyne, 2-propynyloxy benzene, phenylpropargyl ether, prop-2-ynyloxybenzene, ether, phenyl 2-propynyl
PubChem CID 83613
IUPAC Name prop-2-ynoxybenzene
SMILES C#CCOC1=CC=CC=C1

Specifications

Density 1.0300g/mL
Boiling Point 89.0°C to 90.0°C (14.0mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.5340 to 1.5360 (20°C, 589nm)
Linear Formula C6H5OCH2CCH
Quantity 5mL
Specific Gravity 1.03
Formula Weight 132.16
Percent Purity 97+%
Chemical Name or Material Phenyl propargyl ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
EINECSNumber 237-095-5

RUO – Research Use Only