missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Phosphate 99.0+%, TCI America™

Catalog No. B002625G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B002625G 25 g
B0026250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B002625G Supplier TCI America Supplier No. B002625G
Only null left

Chemical Identifiers

CAS 701-64-4
Molecular Formula C6H5Na2O4P
Molecular Weight (g/mol) 218.06
MDL Number MFCD00058970
InChI Key TYJOJLOWRIQYQM-UHFFFAOYSA-L
Synonym Phosphoric Acid Phenyl Ester, Dihydrogen Phenyl Phosphate
PubChem CID 12793
ChEBI CHEBI:37548
IUPAC Name disodium phenyl phosphate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=CC=CC=C1

Specifications

Melting Point 100°C
Quantity 25 g
UN Number 3077
Formula Weight 174.09
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl Phosphate
RTECSNumber SZ9652500
TSCA Yes