missing translation for 'onlineSavingsMsg'
Learn More

Phenyl phosphate disodium salt hydrate, 98%

Catalog No. p-7059492
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4471114 25 g
AA4471122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4471114 Supplier Thermo Scientific Chemicals Supplier No. 04471114
Only null left

Used in phosphatase determinations | CAS: 3279-54-7 | C6H5Na2O4P | 218.06 g/mol

Phenyl phosphate disodium salt is used in phosphatase determinations. It is also used as reagent in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3279-54-7
Molecular Formula C6H5Na2O4P
Molecular Weight (g/mol) 218.06
MDL Number MFCD00002133
InChI Key TYJOJLOWRIQYQM-UHFFFAOYSA-L
Synonym disodium phenyl phosphate, phenyl phosphate disodium salt, disodium phenylphosphate, phenyldisodium phosphate, phosphoric acid, monophenyl ester, disodium salt, phenyl disodium orthophosphate, phenyl phosphate, phosphoric acid, monophenyl ester, sodium salt 1:2, disodium phenyl phosphate dihydrate, phenyl phosphate disodium salt dihydrate
PubChem CID 76771
IUPAC Name disodium;phenyl phosphate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=CC=CC=C1

Specifications

Color Yellow
Linear Formula C6H5OPO(ONa)2·xH2O
Quantity 25 g
Beilstein 4167154
Sensitivity Hygroscopic
Merck Index 14,3357
Solubility Information Soluble in water (0.1g/mL).
Formula Weight 218.06 (Anhydrous)
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Phenyl phosphate disodium salt hydrate
Show More Show Less
EINECSNumber 221-917-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only