missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl beta-D-Galactopyranoside 98.0+%, TCI America™
$109.74 - $317.66
Chemical Identifiers
| CAS | 2818-58-8 |
|---|---|
| Molecular Formula | C12H16O6 |
| Molecular Weight (g/mol) | 256.254 |
| MDL Number | MFCD00063258 |
| InChI Key | NEZJDVYDSZTRFS-YBXAARCKSA-N |
| Synonym | phenylgalactoside, a-d-galactoside, phenyl b-d-galactoside, phenyl b-d-galactoside, phenyl b-d-galactopyranoside, phenyl b-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-phenoxy-tetrahydropyran-3,4,5-triol, p-gal, phenylb-d-galactoside, phenyl, phenyl |
| PubChem CID | 102336 |
| ChEBI | CHEBI:8098 |
| IUPAC Name | (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol |
| SMILES | C1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O |
Chemical Identifiers
| 2818-58-8 | |
| 256.254 | |
| NEZJDVYDSZTRFS-YBXAARCKSA-N | |
| 102336 | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol |
| C12H16O6 | |
| MFCD00063258 | |
| phenylgalactoside, a-d-galactoside, phenyl b-d-galactoside, phenyl b-d-galactoside, phenyl b-d-galactopyranoside, phenyl b-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-phenoxy-tetrahydropyran-3,4,5-triol, p-gal, phenylb-d-galactoside, phenyl, phenyl | |
| CHEBI:8098 | |
| C1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O |
Specifications
| 2818-58-8 | |
| White | |
| MFCD00063258 | |
| phenylgalactoside, a-d-galactoside, phenyl b-d-galactoside, phenyl b-d-galactoside, phenyl b-d-galactopyranoside, phenyl b-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-phenoxy-tetrahydropyran-3,4,5-triol, p-gal, phenylb-d-galactoside, phenyl, phenyl | |
| C1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O | |
| 256.254 | |
| CHEBI:8098 | |
| ≥98.0% (HPLC) | |
| Phenyl beta-D-Galactopyranoside |
| 148°C | |
| C12H16O6 | |
| 1 g | |
| NEZJDVYDSZTRFS-YBXAARCKSA-N | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol | |
| 102336 | |
| 256.25 | |
| Crystalline Powder |
Safety and Handling
TSCA : No