missing translation for 'onlineSavingsMsg'
Learn More

Phenyl beta-D-Galactopyranoside 98.0+%, TCI America™

Catalog No. P13265G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13265G 5 g
P13261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13265G Supplier TCI America Supplier No. P13265G

Chemical Identifiers

CAS 2818-58-8
Molecular Formula C12H16O6
Molecular Weight (g/mol) 256.254
MDL Number MFCD00063258
InChI Key NEZJDVYDSZTRFS-YBXAARCKSA-N
Synonym phenylgalactoside, phenyl beta-d-galactopyranoside, phenyl b-d-galactoside, phenyl-b-d-galactopyranoside, phenyl b-d-galactopyranoside, phenyl-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-phenoxy-tetrahydropyran-3,4,5-triol, p-gal, phenylb-d-galactoside, phenyl, a-d-galactoside
PubChem CID 102336
ChEBI CHEBI:8098
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol
SMILES C1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 148°C
Color White
Quantity 5 g
Formula Weight 256.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl beta-D-Galactopyranoside