missing translation for 'onlineSavingsMsg'
Learn More

Phenyl beta-D-galactopyranoside, 99%

Catalog No. AAH2702803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2702803 1 g
AAH2702806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2702803 Supplier Thermo Scientific Chemicals Supplier No. H2702803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2818-58-8 | C12H16O6 | 256.254 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2818-58-8
Molecular Formula C12H16O6
Molecular Weight (g/mol) 256.254
MDL Number MFCD00063258
InChI Key NEZJDVYDSZTRFS-YBXAARCKSA-N
Synonym phenylgalactoside, phenyl beta-d-galactopyranoside, phenyl b-d-galactoside, phenyl-b-d-galactopyranoside, phenyl b-d-galactopyranoside, phenyl-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-phenoxy-tetrahydropyran-3,4,5-triol, p-gal, phenylb-d-galactoside, phenyl, a-d-galactoside
PubChem CID 102336
ChEBI CHEBI:8098
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol
SMILES C1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 152°C to 156°C
Quantity 1 g
Beilstein 87518
Optical Rotation −42° (c=2 in Water)
Formula Weight 256.25
Percent Purity 99%
Chemical Name or Material Phenyl beta-D-galactopyranoside
EINECSNumber 220-573-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only