missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Benzoate 99.0+%, TCI America™

Catalog No. B007525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B007525G 25 g
B0075100G 100 g
B0075500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B007525G Supplier TCI America Supplier No. B007525G

Chemical Identifiers

CAS 93-99-2
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.221
MDL Number MFCD00003072
InChI Key FCJSHPDYVMKCHI-UHFFFAOYSA-N
Synonym benzoic acid phenyl ester, diphenylcarboxylate, benzoic acid, phenyl ester, benzoic acid phenyl, phenylbenzoate, unii-b8a3wvz590, phenol, benzoate, dsstox_cid_28185, dsstox_rid_82744, dsstox_gsid_48210
PubChem CID 7169
ChEBI CHEBI:86919
IUPAC Name phenyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OC2=CC=CC=C2

Specifications

Melting Point 70°C
Color White
Boiling Point 314°C
Quantity 25 g
Formula Weight 198.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl Benzoate
EINECSNumber (3)-1355
RTECSNumber DH6299500
TSCA Yes