missing translation for 'onlineSavingsMsg'
Learn More

Phenyl benzoate, 99%

Catalog No. p-181868
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162061000 100 g Plastic bottle
AC162065000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC162061000 Supplier Thermo Scientific Chemicals Supplier No. 162061000

CAS: 93-99-2 | C13H10O2 | 198.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93-99-2
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00003072
InChI Key FCJSHPDYVMKCHI-UHFFFAOYSA-N
Synonym benzoic acid phenyl ester, diphenylcarboxylate, benzoic acid, phenyl ester, benzoic acid phenyl, phenylbenzoate, unii-b8a3wvz590, phenol, benzoate, dsstox_cid_28185, dsstox_rid_82744, dsstox_gsid_48210
PubChem CID 7169
ChEBI CHEBI:86919
IUPAC Name phenyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OC2=CC=CC=C2

Specifications

Acidity 1mg KOH/g max.
Melting Point 68.0°C to 70.0°C
Color White
Boiling Point 298.0°C to 299.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5CO2C6H5
Quantity 100 g
Beilstein 09, 116
Merck Index 15, 7388
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether and chloroform
Formula Weight 198.22
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Phenyl benzoate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
EINECSNumber 202-293-2
RTECSNumber DH6299500
TSCA TSCA

RUO – Research Use Only