missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 3,4,6-Tri-O-acetyl-2-deoxy-1-thio-2-(2,2,2-trichloroethoxyformamido)-beta-D-galactopyranoside 98.0+%, TCI America™

Catalog No. p-7135189
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16421G 1 g
P16425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16421G Supplier TCI America Supplier No. P16421G
Only null left

Chemical Identifiers

CAS 278784-83-1
Molecular Formula C21H24Cl3NO9S
Molecular Weight (g/mol) 572.83
MDL Number MFCD11112181,MFCD15072189
InChI Key AZDNCEYOFMNMSH-UHFFFAOYNA-N
PubChem CID 133554192
IUPAC Name [3,4-bis(acetyloxy)-6-(phenylsulfanyl)-5-{[(2,2,2-trichloroethoxy)carbonyl]amino}oxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(SC2=CC=CC=C2)C(NC(=O)OCC(Cl)(Cl)Cl)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 117°C
Color White
Quantity 1 g
Formula Weight 572.83
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl 3,4,6-Tri-O-acetyl-2-deoxy-1-thio-2-(2,2,2-trichloroethoxyformamido)-beta-D-galactopyranoside
TSCA No
Recommended Storage Refrigerator