missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 2,3,4,6-Tetra-O-acetyl-beta-D-thioglucopyranoside, 98%, Thermo Scientific Chemicals

$110.49 - $110.49

Chemical Identifiers

CAS 23661-28-1
Molecular Formula C20H24O9S
Molecular Weight (g/mol) 440.463
MDL Number MFCD00135159
InChI Key JCKOUAWEMPKIAT-OBKDMQGPSA-N
Synonym 2r,3r,4s,5r,6s-2-acetoxymethyl-6-phenylthio tetrahydro-2h-pyran-3,4,5-triyl triacetate, 1-phenyl-1-thio-beta-d-glucopyranoside tetraacetate, phenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-glucopyranoside, phenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-glucopyranoside, 10.14272/jckouawempkiat-obkdmqgpsa-n, 1-phenyl-1-thio-, 1-phenyl-1-thio-, doi:10.14272/jckouawempkiat-obkdmqgpsa-n, doi:10.14272/jckouawempkiat-obkdmqgpsa-n, 1-phenylthio-1-deoxy-beta-d-glucopyranose tetraacetate, 1-phenylthio-1-deoxy-beta-d-glucopyranose tetraacetate
PubChem CID 6364823
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)OC(=O)C)OC(=O)C)OC(=O)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6635503
View Documents
Thermo Scientific Chemicals
J6635503
1 g
Each for $110.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

23661-28-1
440.463
JCKOUAWEMPKIAT-OBKDMQGPSA-N
6364823
CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)OC(=O)C)OC(=O)C)OC(=O)C
C20H24O9S
MFCD00135159
2r,3r,4s,5r,6s-2-acetoxymethyl-6-phenylthio tetrahydro-2h-pyran-3,4,5-triyl triacetate, 1-phenyl-1-thio-beta-d-glucopyranoside tetraacetate, phenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-glucopyranoside, phenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-glucopyranoside, 10.14272/jckouawempkiat-obkdmqgpsa-n, 1-phenyl-1-thio-, 1-phenyl-1-thio-, doi:10.14272/jckouawempkiat-obkdmqgpsa-n, doi:10.14272/jckouawempkiat-obkdmqgpsa-n, 1-phenylthio-1-deoxy-beta-d-glucopyranose tetraacetate, 1-phenylthio-1-deoxy-beta-d-glucopyranose tetraacetate
[(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxan-2-yl]methyl acetate

Specifications

23661-28-1
White
MFCD00135159
2r,3r,4s,5r,6s-2-acetoxymethyl-6-phenylthio tetrahydro-2h-pyran-3,4,5-triyl triacetate, 1-phenyl-1-thio-beta-d-glucopyranoside tetraacetate, phenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-glucopyranoside, phenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-glucopyranoside, 10.14272/jckouawempkiat-obkdmqgpsa-n, 1-phenyl-1-thio-, 1-phenyl-1-thio-, doi:10.14272/jckouawempkiat-obkdmqgpsa-n, doi:10.14272/jckouawempkiat-obkdmqgpsa-n, 1-phenylthio-1-deoxy-beta-d-glucopyranose tetraacetate, 1-phenylthio-1-deoxy-beta-d-glucopyranose tetraacetate
CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)OC(=O)C)OC(=O)C)OC(=O)C
440.463
440.47
Powder
116°C to 119°C
C20H24O9S
1 g
JCKOUAWEMPKIAT-OBKDMQGPSA-N
[(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxan-2-yl]methyl acetate
6364823
98%
Phenyl 2,3,4,6-Tetra-O-acetyl-beta-D-thioglucopyranoside
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only