missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 2,3,4,6-Tetra-O-acetyl-beta-D-thioglucopyranoside, 98%, Thermo Scientific Chemicals

Catalog No. AAJ6635503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6635503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6635503 Supplier Thermo Scientific Chemicals Supplier No. J6635503
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23661-28-1
Molecular Formula C20H24O9S
Molecular Weight (g/mol) 440.463
MDL Number MFCD00135159
InChI Key JCKOUAWEMPKIAT-OBKDMQGPSA-N
Synonym 2r,3r,4s,5r,6s-2-acetoxymethyl-6-phenylthio tetrahydro-2h-pyran-3,4,5-triyl triacetate, phenyl 2,3,4,6-tetra-o-acetyl-b-d-thioglucopyranoside, phenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-glucopyranoside, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-phenylsulfanyl oxan-2-yl methyl acetate, 10.14272/jckouawempkiat-obkdmqgpsa-n, 1-phenyl-1-thio-, a-d-glucopyranoside tetraacetate, doi:10.14272/jckouawempkiat-obkdmqgpsa-n, s-phenyl-1-thio-beta-d-glucopyranoside tetracetate, 1-phenylthio-1-deoxy-beta-d-glucopyranose tetraacetate, 1-phenyl-1-thio-beta-d-glucopyranoside tetraacetate
PubChem CID 6364823
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 116°C to 119°C
Color White
Quantity 1 g
Formula Weight 440.47
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Phenyl 2,3,4,6-Tetra-O-acetyl-beta-D-thioglucopyranoside
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only