missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-beta-D-glucopyranoside 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 118°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 440.46 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-beta-D-glucopyranoside |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |