missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-beta-D-galactopyranoside 98.0+%, TCI America™

Catalog No. p14775g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P14775G Supplier TCI America Supplier No. P14775G

Specifications

CAS 24404-53-3
Melting Point 70°C
Color White
Molecular Formula C20H24O9S
Quantity 5 g
Synonym phenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-galactopyranoside, phenyl 2,3,4,6-tetra-o-acetyl-b-d-thiogalactopyranoside, 2r,3s,4s,5r,6s-2-acetoxymethyl-6-phenylthio tetrahydro-2h-pyran-3,4,5-triyl triacetate, 2r,3s,4s,5r,6s-3,4,5-tris acetyloxy-6-phenylsulfanyl oxan-2-yl methyl acetate, pubchem9897, phenyl2,3,4,6-tetra-o-acetyl-1-thio-beta-d-galactopyranoside, phenyl-tetra-o-acetyl-thio-beta-d-galactopyranoside, 1-phenylthio-1-deoxy-beta-d-galactopyranose tetraacetate, 1-deoxy-1-phenylthio-2,3,4,6-tetra-o-acetyl-beta-d-galactopyranose, phenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-galactopyranoside hplc
InChI Key JCKOUAWEMPKIAT-CXQPBAHBSA-N
SMILES CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)OC(=O)C)OC(=O)C)OC(=O)C
IUPAC Name [(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxan-2-yl]methyl acetate
Molecular Weight (g/mol) 440.463
PubChem CID 10950254
Formula Weight 440.46
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-beta-D-galactopyranoside
Show More Show Less
TSCA No
Recommended Storage Freezer