missing translation for 'onlineSavingsMsg'
Learn More

Phentolamine Mesylate 98.0+%, TCI America™

Catalog No. p-7135348
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1985100MG 100 mg
P19851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1985100MG Supplier TCI America Supplier No. P1985100MG

Chemical Identifiers

CAS 65-28-1
Molecular Formula C18H23N3O4S
Molecular Weight (g/mol) 377.459
MDL Number MFCD00134201
InChI Key OGIYDFVHFQEFKQ-UHFFFAOYSA-N
Synonym phentolamine mesylate, phentolamine mesilate, phentolamine methanesulfonate, oraverse, regitine mesylate, regitin methanesulphonate, regitine methanesulfonate, phentolamine mesylate usp, phentolamine methanesulphonate
PubChem CID 91430
IUPAC Name 3-[N-(4,5-dihydro-1H-imidazol-2-ylmethyl)-4-methylanilino]phenol;methanesulfonic acid
SMILES CC1=CC=C(C=C1)N(CC2=NCCN2)C3=CC(=CC=C3)O.CS(=O)(=O)O

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 100 mg
Formula Weight 377.46
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phentolamine Mesylate
RTECSNumber SL5430000
TSCA No