missing translation for 'onlineSavingsMsg'
Learn More

Phenoxazine 98.0+%, TCI America™

Catalog No. p-7134834
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08991G 1 g
P08995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08991G Supplier TCI America Supplier No. P08991G
Only null left

Chemical Identifiers

CAS 135-67-1
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00005014
InChI Key TZMSYXZUNZXBOL-UHFFFAOYSA-N
Synonym phenoxazine, phenazoxine, unii-c2zwt499sg, c2zwt499sg, 5,4-oxazine, phenotazine, rarechem fh 2c 0s92, phenoxazine 1g, 9ci
PubChem CID 67278
IUPAC Name 10H-phenoxazine
SMILES N1C2=CC=CC=C2OC2=CC=CC=C12

Specifications

Melting Point 154°C
Quantity 1 g
Formula Weight 183.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenoxazine